Please login to your online account to display your discounted pricing

Thymol Blue Sodium Salt, ACS Grade, LabChem™

$66.37 - $66.37

Chemical Identifiers

Chemical Name or Material Thymol Blue Sodium Salt
CAS 62625-21-2
CAS Max % 100
Purity Grade ACS Grade
Linear Formula C27H29NaO5S
Formula Weight 488.58
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

LC260157

 
lab chem
LC260157
Amber Glass 5g Each for $66.37
Description & Specifications

Specifications

Formula Weight 488.58
Quantity 5g
Identification Passes Test
Chemical Name or Material Thymol Blue Sodium Salt
Purity Grade ACS Grade
Recommended Storage Room Temperature
Linear Formula C27H29NaO5S
Health Hazard 2 GHS H Statement
Product is not hazardous.
Color Dark greenish gray
Physical Form Powder
Solubility Soluble in water
CAS 62625-21-2
Packaging Amber Glass
CAS Max % 100
Health Hazard 3 GHS P Statement
Wear eye protection, protective gloves.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing.
If skin irritation occurs: Get medical advice/attention.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Decomposition Information Sulfur compounds; Carbon monoxide; Carbon dioxide
Safety and Handling

GHS P Statement
Wear eye protection, protective gloves.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing.
If skin irritation occurs: Get medical advice/attention.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
Product is not hazardous.

Room Temperature