missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Thymol Blue sodium salt, 0.04% w/v aq. soln., Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 038692AE
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Thymol Blue sodium salt | |
0.04% w/v aq. soln. | |
MFCD00151093 | |
[Na+].CC(C)C1=C\C(=C(\C2=CC(C(C)C)=C(O)C=C2C)C2=C(C=CC=C2)S([O-])(=O)=O)C(C)=CC1=O | |
488.57 | |
488.58 | |
0.979 g/mL | |
Liquid |
62625-21-2 | |
C27H29NaO5S | |
OCMIKNBSIRSUPI-FRWNMSGJSA-M | |
sodium;4-[3-(4-hydroxy-2-methyl-5-propan-2-ylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-5-methyl-2-propan-2-ylphenolate | |
23692293 | |
Odorless | |
100 mL |
Chemical Identifiers
62625-21-2 | |
488.57 | |
OCMIKNBSIRSUPI-FRWNMSGJSA-M | |
sodium;4-[3-(4-hydroxy-2-methyl-5-propan-2-ylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-5-methyl-2-propan-2-ylphenolate |
C27H29NaO5S | |
MFCD00151093 | |
23692293 | |
[Na+].CC(C)C1=C\C(=C(\C2=CC(C(C)C)=C(O)C=C2C)C2=C(C=CC=C2)S([O-])(=O)=O)C(C)=CC1=O |
Safety and Handling
EINECSNumber : 200-973-3
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only