Please login to your online account to display your discounted pricing

Thymol Blue Indicator, 0.04% (w/v) in Alcohol, Ricca Chemical


Quantity 1L
Packaging Plastic Bottle

Manufacturer: ricca chemical company  RIR84210001A

 View more versions of this product

Catalog No. R84210001A

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Liquid
Color Clear dark red
Quantity 1L
Chemical Name or Material Thymol Blue Indicator
CAS 64-17-5
Packaging Plastic Bottle
CAS Min % 94.0
CAS Max % 6.0
CAS Min % 4.0
CAS Max % 0.05
CAS Min % 0.05
CAS Max % 96.0
CAS 76-61-9
CAS 67-56-1

Specifications

Chemical Name or Material Thymol Blue Indicator
CAS 64-17-5
CAS Min % 94.0
CAS Max % 96.0
CAS 76-61-9
CAS Min % 0.05
CAS Max % 0.05
CAS 67-56-1
CAS Min % 4.0
CAS Max % 6.0
Physical Form Liquid