Please login to your online account to display your discounted pricing

Thymol Blue, 0.08% in Methanol, for Organic Acids, Certified, LabChem™

$67.46 - $67.46

Chemical Identifiers

Chemical Name or Material Thymol Blue
CAS 67-56-1
CAS Max % 99.92
CAS 76-16-9
CAS Max % 0.08
Name Note 0.08% in Methanol, for Organic Acids
Purity Grade Certified Grade
Linear Formula C27H29NaO5S
Formula Weight 488.58
UN Number UN1993
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

LC260501

 
lab chem
LC260501
Poly Bottle 500mL Each for $67.46
Description & Specifications

Specifications

Purity Grade Certified Grade
Formula Weight 488.58
CAS Max % 99.92
Boiling Point 65°C
CAS Max % 0.08
Linear Formula C27H29NaO5S
Identification Passes Test
Health Hazard 3 GHS P Statement
Wear eye protection, protective gloves.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing.
If skin irritation occurs: Get medical advice/attention.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
CAS 67-56-1
Health Hazard 1 Danger
Packaging Poly Bottle
Physical Form Liquid
Solubility Soluble in water
Specific Gravity 0.70g/mL
Decomposition Information Sulfur compounds; Carbon monoxide; Carbon dioxide; May release flammable gases
Quantity 500mL
Melting Point −98°C
Flash Point 9.7°C
Color Deep red
UN Number UN1993
Health Hazard 2 GHS H Statement
Highly flammable liquid and vapor.
Toxic if swallowed, in contact with skin or if inhaled.
Causes damage to organs (central nervous system, kidneys, liver, optic nerve).
CAS 76-16-9
Density 0.70g/mL
Recommended Storage Refrigerate
Name Note 0.08% in Methanol, for Organic Acids
Chemical Name or Material Thymol Blue
Safety and Handling

Danger

GHS P Statement
Wear eye protection, protective gloves.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing.
If skin irritation occurs: Get medical advice/attention.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
Highly flammable liquid and vapor.
Toxic if swallowed, in contact with skin or if inhaled.
Causes damage to organs (central nervous system, kidneys, liver, optic nerve).

Refrigerate