Please login to your online account to display your discounted pricing

Thymol Blue, 0.04% Aqueous, pH 1.2 to 2.8 Red to Yellow, pH 8.0 to 9.6 Yellow to Blue, Certified, LabChem™

$25.74 - $25.74

Chemical Identifiers

Chemical Name or Material Thymol Blue
CAS 7732-18-5
CAS Max % 99.96
CAS 62625-21-2
CAS Max % 0.04
Name Note 0.04% Aqueous, pH 1.2 to 2.8 Red to Yellow, pH 8.0 to 9.6 Yellow to Blue
Purity Grade Certified Grade
Linear Formula C27H29NaO5S
Formula Weight 488.58
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

LC260207

 
lab chem
LC260207
Poly Bottle 125mL Each for $25.74
Description & Specifications

Specifications

Purity Grade Certified Grade
Formula Weight 488.58
Name Note 0.04% Aqueous, pH 1.2 to 2.8 Red to Yellow, pH 8.0 to 9.6 Yellow to Blue
Linear Formula C27H29NaO5S
CAS 7732-18-5
Identification Passes Test
Packaging Poly Bottle
CAS Max % 99.96
CAS 62625-21-2
Health Hazard 3 GHS P Statement
Wear eye protection, protective gloves.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing.
If skin irritation occurs: Get medical advice/attention.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Solubility Soluble in water
Recommended Storage Room Temperature
Decomposition Information Sulfur compounds; Carbon monoxide; Carbon dioxide
Color Amber
Chemical Name or Material Thymol Blue
Quantity 125mL
Physical Form Liquid
Health Hazard 2 GHS H Statement
Solution is not hazardous.
CAS Max % 0.04
Safety and Handling

GHS P Statement
Wear eye protection, protective gloves.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing.
If skin irritation occurs: Get medical advice/attention.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
Solution is not hazardous.

Room Temperature