missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thymol, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 150331000

 View more versions of this product

Catalog No. AC150331000



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Thymol
89-83-8
White
232.0°C
Authentic
Plastic bottle
MFCD00002309
06, 532
0.96
thymol, 2-isopropyl-5-methylphenol, 5-methyl-2-isopropylphenol, thyme camphor, 3-p-cymenol, 6-isopropyl-m-cresol, thymic acid, isopropyl cresol, 5-methyl-2-1-methylethyl phenol, 6-isopropyl-3-methylphenol
MGSRCZKZVOBKFT-UHFFFAOYSA-N
5-methyl-2-propan-2-ylphenol
6989
CHEBI:27607
99%
99%
48.0°C to 52.0°C
0.9600g/mL
102°C
98.5% min. (GC)
C10H14O
100 g
0.05% max.
15, 9554
Solubility in water: 0.1% (20°C). Other solubilities: soluble in glacial acetic acid, soluble in alcohol, carbon disulfide, chloroform, and ether, slightly soluble in glycerol
CC1=CC(=C(C=C1)C(C)C)O
150.22
4.3 mm2/s (50°C)
150.22
Crystalline Powder or Crystals

Chemical Identifiers

89-83-8
150.22
MGSRCZKZVOBKFT-UHFFFAOYSA-N
6989
5-methyl-2-propan-2-ylphenol
C10H14O
MFCD00002309
thymol, 2-isopropyl-5-methylphenol, 5-methyl-2-isopropylphenol, thyme camphor, 3-p-cymenol, 6-isopropyl-m-cresol, thymic acid, isopropyl cresol, 5-methyl-2-1-methylethyl phenol, 6-isopropyl-3-methylphenol
CHEBI:27607
CC1=CC(=C(C=C1)C(C)C)O
Safety and Handling

Safety and Handling

GHS H Statement:
Toxic to aquatic life with long lasting effects.
Harmful if swallowed.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 201-944-8

RTECSNumber : XP2275000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only