missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Thymidine Phosphorylase/PD-ECGF Ab-1, Mouse Monoclonal Antibody

Provide accurate, reproducible results with the Thermo Scientific™ Thymidine Phosphorylase/PD-ECGF Ab-1, Mouse Monoclonal Antibody.

$334.51 - $1,720.80

Specifications

Antigen Thymidine Phosphorylase/PD-ECGF Ab-1
Clone P-GF.44C
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 6
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
MS499P View Documents Thermo Scientific™
MS499P
Purified Ab with BSA and Azide 1mL Each for $1,597.04
 
MS499P0 View Documents Thermo Scientific™
MS499P0
Purified Ab with BSA and Azide 100μL Each for $334.51
 
MS499P1 View Documents Thermo Scientific™
MS499P1
Purified Ab with BSA and Azide 500μL Each for $948.06
 
MS499P1ABX View Documents Thermo Scientific™
MS499P1ABX
Ab without BSA/Azide (Frozen) 100μL Each for $1,052.78
 
MS499PABX View Documents Thermo Scientific™
MS499PABX
Ab without BSA/Azide (Frozen) 200μL Each for $1,720.80
 
MS499R7 View Documents Thermo Scientific™
MS499R7
Ready-to-use for IHC (UVLP) 7mL Each for $819.82
 
Description

Description

Provide accurate, reproducible results with the Thermo Scientific™ Thymidine Phosphorylase/PD-ECGF Ab-1, Mouse Monoclonal Antibody.

Platelet-derived endothelial growth factor (PD-ECGF), same as thymidine phosphorylase (TP) or gliostatin. In the presence of inorganic orthophosphate, it catalyses the reversible phospholytic cleavage of thymidine and deoxyuridine to their corresponding bases and 2-deoxyribose-1-phosphate. It is both chemotactic and mitogenic for endothelial cells and a non-heparin binding angiogenic factor present in platelets. It is also involved in transformation of fluoropyrimidines, cytotoxic agents used in the treatment of a variety of malignancies, into active cytotoxic metabolites.

Host Species: Mouse

Clone: P-GF.44C

Isotype: IgG1

Species Reactivity: Human, Mouse and Rat. Others not known.

Epitope: Not determined

Immunogen: Recombinant full-length human thymidine phosphorylase (TP/PD-ECGF) protein

Molecular Weight: 55kDa (reduced)

Positive Control: HUVEC cells, Breast CA

Cellular Localization: Cytoplasmic and nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting (Not verified)
  • Neutralizes Enzymatic Activity of TP/PD-ECGF (Not verified)

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

Thymidine Phosphorylase/PD-ECGF Ab-1
Immunohistochemistry (Paraffin)
Unconjugated
RUO
Primary
IgG1
P-GF.44C
Monoclonal
Murine
Angiogenesis
Recombinant full length human thymidine phosphorylase (TP/PD-ECGF) protein