missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Thymidine, 99%

Manufacturer:  Alfa Aesar™ A1149314

 View more versions of this product

Catalog No. AAA1149314



Description

Description

Applications
Thymidine is used in the syntheses of active pharmaceutical ingredient such as zidovudine. It also pairs with deoxyadenosine in double-stranded deoxyribonucleic acid. It is used to synchronize the cells in G1/early S phase in cell biology.

Solubility
Soluble in water, methanol, hot ethanol, hot acetone, hot ethyl acetate, pyridine, glacial acetic acid and hydrochloric acid. Slightly soluble in hot chloroform .

Notes
Store in a cool place. Incompatible with strong oxidizing agents.
Specifications

Specifications

Thymidine
+19° (c=1 in water)
25g
C10H14N2O5
89285
thymidine, deoxythymidine, 2'-deoxythymidine, 5-methyldeoxyuridine, thymidin, beta-thymidine, dthyd, 5-methyl-2'-deoxyuridine, thymine-2-deoxyriboside, thyminedeoxyriboside
CC1=CN(C(=O)NC1=O)C2CC(C(O2)CO)O
242.231
CHEBI:17748
Powder
Odorless
186°C to 190°C
50-89-5
MFCD00006537
14,9397
IQFYYKKMVGJFEH-XLPZGREQSA-N
1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-methylpyrimidine-2,4-dione
5789
242.23
99%

Chemical Identifiers

50-89-5
242.231
IQFYYKKMVGJFEH-XLPZGREQSA-N
5789
1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-methylpyrimidine-2,4-dione
C10H14N2O5
MFCD00006537
thymidine, deoxythymidine, 2'-deoxythymidine, 5-methyldeoxyuridine, thymidin, beta-thymidine, dthyd, 5-methyl-2'-deoxyuridine, thymine-2-deoxyriboside, thyminedeoxyriboside
CHEBI:17748
CC1=CN(C(=O)NC1=O)C2CC(C(O2)CO)O
Safety and Handling

Safety and Handling

RTECSNumber : XP2071000

TSCA : TSCA

SDS Documents