Please login to your online account to display your discounted pricing
Novus Biologicals™ Thunder-Link oligo Antibody Labeling Kit
Antibody-oligonucleotide conjugates are essential to immuno-PCR. The Thunder-Link™ oligo conjugation system can generate these conjugates more quickly and easily than standard chemistry and methodology.
$483.00 - $706.00
Products
Catalog Number Mfr. No. Quantity Price Quantity    
4200000
novus biologicals™
420-0000
1 Reaction Each for $483.00
4200300
novus biologicals™
420-0300
3 Reactions Each for $706.00
Description & Specifications
Features
  • End-user chooses oligo sequence and antibody
  • Oligo sizes (HPLC purified): 10 to 120 bases
  • Amine group positioned at the 5' or 3'
  • Positive control included
  • All necessary components for a successful conjugation are supplied
  • Ideal for immuno-PCR