missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Thulium powder, -40 mesh, 99.9% (REO)

Manufacturer:  Alfa Aesar™ 4416806

 View more versions of this product

Catalog No. AA4416806



Description

Description

  • Packaged under Argon
Specifications

Specifications

Thulium powder
7440-30-4
Tm
UN3089
8rkc5ati4p
[Tm]
Odorless
23961
Powder
Air and moisture sensitive
5 g
(REO)
MFCD00011281
−40 Mesh
FRNOGLGSGLTDKL-UHFFFAOYSA-N
thulium
168.934
CHEBI:33380
99.9%
Gray to Silver

Chemical Identifiers

7440-30-4
168.934
FRNOGLGSGLTDKL-UHFFFAOYSA-N
23961
thulium
Tm
MFCD00011281
8rkc5ati4p
CHEBI:33380
[Tm]
Safety and Handling

Safety and Handling

GHS H Statement
H228-H260
Flammable solid.
In contact with water releases flammable gases which may ignite spontaneously.

P210-P240-P241-P280-P370+P378q

H228-H500

DOTInformation : Hazard Class: 4.1; Packaging Group: III

EINECSNumber : 231-140-2

TSCA : Yes

storageNote1 :

  • Packaged under Argon

Recommended Storage : Ambient temperatures; Store under Argon

Documents