missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Thulium pieces, sublimed dendritic, 99.9% (REO)

Manufacturer:  Alfa Aesar™ 1243506

 View more versions of this product

Catalog No. AA1243506



Description

Description

  • Packaged under argon
Specifications

Specifications

Thulium pieces
Sublimed dendritic
Odorless
Tm
UN3178
FRNOGLGSGLTDKL-UHFFFAOYSA-N
thulium
23961
Pieces
5g
7440-30-4
(REO)
MFCD00011281
tulio, unii-8rkc5ati4p, 8rkc5ati4p, atom, pieces, powder, foil, acmc-20ajig, thulium, ingot, pieces, sublimed dendritic
[Tm]
168.934
CHEBI:33380
99.9%

Chemical Identifiers

7440-30-4
168.934
FRNOGLGSGLTDKL-UHFFFAOYSA-N
23961
thulium
Tm
MFCD00011281
tulio, unii-8rkc5ati4p, 8rkc5ati4p, atom, pieces, powder, foil, acmc-20ajig, thulium, ingot, pieces, sublimed dendritic
CHEBI:33380
[Tm]
Safety and Handling

Safety and Handling

GHS H Statement
H228-H260
Flammable solid.
In contact with water releases flammable gases which may ignite spontaneously.

GHS P Statement
P210-P231+P232-P280-P370+P378c-P402+P404-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Handle under inert gas.
Protect from moisture.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, sand, extinguishing powder.
Store in a dry place.
Store in a closed container.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: FLAMMABLE SOLID, INORGANIC, N.O.S.

TSCA : TSCA

storageNote1 :

  • Packaged under argon

Documents