Learn More
Thulium(III) trifluoromethanesulfonate, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 04031309
Description
A raw material used in synthesis of chemicals .
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Thulium(III) trifluoromethanesulfonate | |
Powder | |
C3F9O9S3Tm | |
MFCD00209617 | |
Soluble in water. | |
C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].[Tm+3] | |
616.124 | |
616.13 |
141478-68-4 | |
10 g | |
Tm(OSO2CF3)3 | |
thulium iii trifluoromethanesulfonate, thulium iii triflate, trifluoromethanesulfonic acid thulium iii salt, thulium triflate, tm otf 3, acmc-1c8m6, thulium 3+ tritriflate, thulium 3+ ; trifluoromethanesulfonate, thulium trifluoromethanesulfonate, thulium iii tris trifluoromethanesulfonate | |
PBASUZORNBYVFM-UHFFFAOYSA-K | |
thulium(3+);trifluoromethanesulfonate | |
2733936 | |
98% |
Chemical Identifiers
141478-68-4 | |
616.124 | |
PBASUZORNBYVFM-UHFFFAOYSA-K | |
2733936 | |
C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].[Tm+3] |
C3F9O9S3Tm | |
MFCD00209617 | |
thulium iii trifluoromethanesulfonate, thulium iii triflate, trifluoromethanesulfonic acid thulium iii salt, thulium triflate, tm otf 3, acmc-1c8m6, thulium 3+ tritriflate, thulium 3+ ; trifluoromethanesulfonate, thulium trifluoromethanesulfonate, thulium iii tris trifluoromethanesulfonate | |
thulium(3+);trifluoromethanesulfonate |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314
DOTInformation : Hazard Class: 8; Packaging Group: II
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only