Please login to your online account to display your discounted pricing

Alfa Aesar™ Thulium(III) sulfate octahydrate, 99.99%

$66.19 - $66.19

Chemical Identifiers

Chemical Name or Material Thulium(III) sulfate octahydrate
CAS 13778-40-0
Molecular Formula Tm2(SO4)3·8H2O
Formula Weight 770.17 (626.06 Anhydrous)
MDL Number MFCD00149881
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1729603

 
alfa aesar™
A1729603
1g Each for $66.19
Description & Specifications

Specifications

Formula Weight 770.17 (626.06 Anhydrous)
Chemical Name or Material Thulium(III) sulfate octahydrate
TSCA TSCA
Assay 99.99%
Molecular Formula Tm2(SO4)3·8H2O
MDL Number MFCD00149881
CAS 13778-40-0
Quantity 1g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Applications
It is used as a reactant in dehydration reactions. It is the important dopant for fiber amplifiers.

Solubility
Soluble in water, moderately soluble in strong mineral acids.

Notes
Store at room temperature. Keep away from strong oxidizing agents and moisture.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.