Please login to your online account to display your discounted pricing

Thulium(III) oxide, 99.99% (REO), REacton™, Alfa Aesar™

Chemical Identifiers

CAS 12036-44-1
Physical Form Powder
Assay Percent Range 99.99% (REO)
Molecular Formula O3Tm2
Formula Weight 385.87
MDL Number MFCD00011285
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1119904

  SDS
alfa aesar
1119904
2g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1119909

  SDS
alfa aesar
1119909
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 385.87
Quantity 2g
Color Pale green
Physical Form Powder
MDL Number MFCD00011285
CAS 12036-44-1
Assay Percent Range 99.99% (REO)
Molecular Formula O3Tm2