missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Thulium(III) oxalate hydrate, REacton™, 99.9% (REO)

Manufacturer:  Alfa Aesar™ 1457804

 View more versions of this product

Catalog No. AA1457804



Specifications

Specifications

Thulium(III) oxalate hydrate
(REO)
Tm2(C2O4)3·xH2O
UN3288
thulium oxalate hydrate, thulium iii oxalate hydrate, acmc-20ajie, ethanedioic acid,thulium 3+ salt,hydrate 3:2:?
DUELVSSHRTZSBY-UHFFFAOYSA-H
oxalate;thulium(3+);hydrate
45052048
Crystalline
Reacton™
58176-73-1
C6H2O13Tm2
MFCD00050804
14,9394
Soluble in aqueous alkali oxalates with double oxalate formation
C(=O)(C(=O)[O-])[O-].C(=O)(C(=O)[O-])[O-].C(=O)(C(=O)[O-])[O-].O.[Tm+3].[Tm+3]
619.937
601.93 (Anhydrous)
99.9%
2g

Chemical Identifiers

58176-73-1
619.937
DUELVSSHRTZSBY-UHFFFAOYSA-H
45052048
C(=O)(C(=O)[O-])[O-].C(=O)(C(=O)[O-])[O-].C(=O)(C(=O)[O-])[O-].O.[Tm+3].[Tm+3]
C6H2O13Tm2
MFCD00050804
thulium oxalate hydrate, thulium iii oxalate hydrate, acmc-20ajie, ethanedioic acid,thulium 3+ salt,hydrate 3:2:?
oxalate;thulium(3+);hydrate
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement
P280-P301+P312-P312-P363-P322-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wash contaminated clothing before reuse.
Specific measures (see label).
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.

TSCA : TSCA

SDS Documents