Please login to your online account to display your discounted pricing

Alfa Aesar™ Thulium(III) iodide, ultra dry, 99.99% (REO)

Chemical Identifiers

Chemical Name or Material Thulium(III) iodide
CAS 13813-43-9
Name Note Ultra dry
Linear Formula TmI3
Formula Weight 549.65
MDL Number MFCD00050141
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3942403

  SDS
alfa aesar™
3942403
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3942406

  SDS
alfa aesar™
3942406
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 549.65
Packaging Ampule
Chemical Name or Material Thulium(III) iodide
TSCA TSCA
Name Note Ultra dry
Physical Form Flakes
Assay 99.99%
Sensitivity Hygroscopic
Assay Percent Notes (REO)
Linear Formula TmI3
MDL Number MFCD00050141
CAS 13813-43-9
Boiling Point 1260°C
Melting Point 1015°C
Quantity 1g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Storage Note 1
  • Ampouled under argon

  • Ampouled under argon

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

  • Ampouled under argon