missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thulium(III) chloride hydrate, 99.99% (REO), REacton™, Alfa Aesar™

Manufacturer:  Thermo Fisher Scientific 1120102

 View more versions of this product

Catalog No. AA1120102



Specifications

Specifications

19423-86-0
TmCl3·xH2O
275.29
Odorless
0.5g
99.99% (REO)
MFCD00011283
Wet Crystalline Aggregates
Green

Chemical Identifiers

19423-86-0
TmCl3·xH2O
MFCD00011283
Documents