missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thulium(III) chloride hydrate, 99.99% (REO), REacton™, Alfa Aesar™

Manufacturer:  Thermo Fisher Scientific 1120109

 View more versions of this product

Catalog No. AA1120109



Specifications

Specifications

19423-86-0
TmCl3·xH2O
Soluble in water.
Wet Crystalline Aggregates
Hygroscopic
10g
99.99% (REO)
MFCD00011283
275.29 (Anhydrous)
Odorless
Green

Chemical Identifiers

19423-86-0
TmCl3·xH2O
MFCD00011283
Safety and Handling

Safety and Handling

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 236-904-9

RTECSNumber : XP0525000

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

Documents