missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thulium(III) chloride hydrate, 99.9% (REO), REacton™, Alfa Aesar™

Chemical Identifiers

CAS 19423-86-0
Physical Form Crystalline Aggregates
Assay Percent Range 99.9% (REO)
Molecular Formula Cl3Tm·xH2O
Formula Weight 275.29
MDL Number MFCD00011283
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA1120004 SDS Alfa Aesar
1120004
2g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AA1120009 SDS Alfa Aesar
1120009
10g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

19423-86-0
99.9% (REO)
275.29
Crystalline Aggregates
Cl3Tm·xH2O
MFCD00011283

Specifications

19423-86-0
99.9% (REO)
275.29
Odorless
2g
Crystalline Aggregates
Cl3Tm·xH2O
MFCD00011283
Green