Learn More
Thulium(III) bromide, ultra dry, 99.99% (REO), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 04719606
Description
Thulium(III) bromide is used for development of advanced ceramic discharge lamps1 and metal halide lamp. Reagent for complexation of lanthanide bromides with aluminum bromide. Reactant for preparation of alkali metal thulium bromides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
1440°C | |
952°C | |
Beads | |
Br3Tm | |
thulium bromide, thulium tribromide, thulium iii bromide, thulium 3+ tribromide, acmc-1bo43, thulium iii bromide, anhydrous, ampuled under argon trace metals basis 1g | |
HQSWGSFQSCMHFQ-UHFFFAOYSA-K | |
thulium(3+);tribromide | |
84454 | |
99.99% | |
Hygroscopic |
Thulium(III) bromide | |
14456-51-0 | |
5 g | |
MFCD00050138 | |
Soluble in water. | |
[Br-].[Br-].[Br-].[Tm+3] | |
408.646 | |
408.66 | |
Plastic bottle; Ampouled under argon |
Chemical Identifiers
14456-51-0 | |
408.646 | |
HQSWGSFQSCMHFQ-UHFFFAOYSA-K | |
84454 | |
[Br-].[Br-].[Br-].[Tm+3] |
Br3Tm | |
MFCD00050138 | |
thulium bromide, thulium tribromide, thulium iii bromide, thulium 3+ tribromide, acmc-1bo43, thulium iii bromide, anhydrous, ampuled under argon trace metals basis 1g | |
thulium(3+);tribromide |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 238-444-4
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only