Learn More
Thulium(III) acetate hydrate, REacton™, 99.9% (REO), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 01458209
Description
Thulium Acetate is the important dopant for silica-based fiber amplifiers. It can also be used in portable X-ray devices that has been bombarded in a nuclear reactor as a radiation source. Used as laboratory reagent, optical glasses, structural ceramics, catalysts, electrical components and photo-optical material. The other main application is used for optical fiber.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Thulium(III) acetate hydrate | |
Crystalline Aggregates | |
(REO) | |
MFCD00013046 | |
Moderately soluble in water,strong mineral acids. | |
[Tm+3].CC([O-])=O.CC([O-])=O.CC([O-])=O | |
346.07 | |
99.9% | |
Hygroscopic |
314041-04-8 | |
10 g | |
C6H9O6Tm | |
acmc-1afih, thulium acetate-water 1/3/1, thulium 3+ ion triacetate hydrate | |
SNMVVAHJCCXTQR-UHFFFAOYSA-K | |
thulium(3+) triacetate | |
346.07 (Anhydrous) | |
Reacton™ |
Chemical Identifiers
314041-04-8 | |
346.07 | |
SNMVVAHJCCXTQR-UHFFFAOYSA-K | |
thulium(3+) triacetate |
C6H9O6Tm | |
MFCD00013046 | |
acmc-1afih, thulium acetate-water 1/3/1, thulium 3+ ion triacetate hydrate | |
[Tm+3].CC([O-])=O.CC([O-])=O.CC([O-])=O |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 254-326-5
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only