Please login to your online account to display your discounted pricing

Thulium(III) 2,4-pentanedionate, Powder, REacton™, 99.9% (REO), Alfa Aesar™

Chemical Identifiers

Chemical Name or Material Thulium(III) 2, 4-pentanedionate
CAS 14589-44-7
Physical Form Powder
Assay Percent Range 99.9% (REO)
Molecular Formula C15H21O6Tm
Formula Weight 466.26
MDL Number MFCD00058840
Synonym Thulium(III) acetylacetonate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1316403

 
alfa aesar
1316403
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1316406

 
alfa aesar
1316406
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 466.26
Quantity 1g
Chemical Name or Material Thulium(III) 2, 4-pentanedionate
Odor Odorless
Physical Form Powder
Assay Percent Range 99.9% (REO)
Molecular Formula C15H21O6Tm
MDL Number MFCD00058840
CAS 14589-44-7
Synonym Thulium(III) acetylacetonate