missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Thulium foil, 0.25mm (0.01 in.) thick, 99.9% (REO)

Manufacturer:  Alfa Aesar™ 10375FF

 View more versions of this product

Catalog No. AA10375FF



Description

Description

  • Packaged under argon
Specifications

Specifications

Thulium foil
25 x 25 mm
7440-30-4
Tm
UN3178
FRNOGLGSGLTDKL-UHFFFAOYSA-N
thulium
168.934
CHEBI:33380
99.9%
≈1.75g/25x25mm
0.25mm (0.01 in.) thick
(REO)
MFCD00011281
8rkc5ati4p, acmc-20ajig, atom, foil, pieces, pieces, sublimed dendritic, powder, thulium, ingot, tulio, unii-8rkc5ati4p
[Tm]
Odorless
23961
Foil
Silver to Gray

Chemical Identifiers

7440-30-4
168.934
FRNOGLGSGLTDKL-UHFFFAOYSA-N
23961
thulium
Tm
MFCD00011281
8rkc5ati4p, acmc-20ajig, atom, foil, pieces, pieces, sublimed dendritic, powder, thulium, ingot, tulio, unii-8rkc5ati4p
CHEBI:33380
[Tm]
Safety and Handling

Safety and Handling

GHS H Statement
H228-H260
Flammable solid.
In contact with water releases flammable gases which may ignite spontaneously.

P210-P223-P231+P232-P240-P241-P280-P335+P334-P370+P378q-P501c

H228-H260

DOTInformation : Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: FLAMMABLE SOLID, INORGANIC, N.O.S.

EINECSNumber : 231-140-2

TSCA : Yes

storageNote1 :

  • Packaged under argon

Recommended Storage : Ambient temperatures

Documents