missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Thrombospondin (TSP) Ab-4, Mouse Monoclonal Antibody

Provide accurate, reproducible results Thermo Scientific™ Thrombospondin (TSP) Ab-4, Mouse Monoclonal Antibody.

$341.86 - $1,993.12

Specifications

Antigen Thrombospondin (TSP) Ab-4
Clone A6.1
Applications Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Host Species Murine
View More Specs

Products 9
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
MS421B View Documents Thermo Scientific™
MS421B
Biotinylated Ab with BSA and Azide 1mL Each for $1,993.12
 
MS421B0 View Documents Thermo Scientific™
MS421B0
Biotinylated Ab with BSA and Azide 100μL Each for $456.69
 
MS421B1 View Documents Thermo Scientific™
MS421B1
Biotinylated Ab with BSA and Azide 500μL Each for $1,259.94
 
MS421P View Documents Thermo Scientific™
MS421P
Purified Ab with BSA and Azide 1mL Each for $1,615.31
 
MS421P0 View Documents Thermo Scientific™
MS421P0
Purified Ab with BSA and Azide 100μL Each for $341.86
 
MS421P1 View Documents Thermo Scientific™
MS421P1
Purified Ab with BSA and Azide 500μL Each for $962.29
 
MS421P1ABX View Documents Thermo Scientific™
MS421P1ABX
Ab without BSA/Azide (Frozen) 100μL Each for $1,058.08
 
MS421PABX View Documents Thermo Scientific™
MS421PABX
Ab without BSA/Azide (Frozen) 200μL Each for $1,707.61
 
MS421R7 View Documents Thermo Scientific™
MS421R7
Ready-to-use for IHC (UVLP) 7mL Each for $1,039.73
 
Description

Description

Provide accurate, reproducible results Thermo Scientific™ Thrombospondin (TSP) Ab-4, Mouse Monoclonal Antibody.

Thrombospondin is a protein from platelet a-granules. It is secreted at sites of platelet activation and aggregation and is involved in the processes of chemotaxis, adhesion, proliferation and differentiation of leukocytes, fibroblasts, smooth muscle and endothelial cells.

Host Species: Mouse

Clone: A6.1

Isotype: IgG1

Species Reactivity: Human, Cow, Pig, Horse, Dog, Sheep, Mouse and Rat. Others not known.

Epitope: Collagen Type V-binding domain of TSP

Immunogen: Reduced and alkylated purified human TSP (fully denatured) from the supernatant of thrombin-activated platelets

Molecular Weight: ∽450kDa (nonreduced), 170 to 180kDa (reduced)

Positive Control: Tonsil or Placenta

Cellular Localization: Secretory granules, Golgi complex, endoplasmic reticulum, extracellular matrix

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Native verified)
  • Electron Microscopy
  • Flow Cytometry

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

Thrombospondin (TSP) Ab-4
Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Murine
Angiogenesis
Reduced and alkylated purified human TSP (fully denatured) from the supernatant of thrombin-activated platelets
A6.1
Monoclonal
RUO
Primary
IgG1