Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Thrombospondin (TSP) Ab-3, Mouse Monoclonal Antibody

Provide accurate, reproducible results with the Thermo Scientific™ Thrombospondin (TSP) Ab-3, Mouse Monoclonal Antibody.

$262.47 - $1,324.76

Specifications

Antigen Thrombospondin (TSP) Ab-3
Clone C6.7
Applications Flow Cytometry
Applications Immunofluorescence
Applications Immunoprecipitation
View More Specs
Products
Catalog Number Mfr. No. Concentration Quantity Price Quantity    

MS420P

 
thermo scientific™ lab vision™
MS420P
200μg/mL 1mL Each for $1,231.61

MS420P0

 
thermo scientific™ lab vision™
MS420P0
200μg/mL 100µL Each for $262.47

MS420P1

 
thermo scientific™ lab vision™
MS420P1
200μg/mL 500µL Each for $735.53

MS420P1ABX

 
thermo scientific™ lab vision™
MS420P1ABX
1mg/mL 100µL Each for $813.73

MS420PABX

 
thermo scientific™ lab vision™
MS420PABX
1mg/mL 200µL Each for $1,324.76
Description & Specifications

Specifications

Antigen Thrombospondin (TSP) Ab-3
Clone C6.7
Applications Flow Cytometry
Applications Immunofluorescence
Applications Immunoprecipitation
Applications Western Blotting
Applications Flow Cytometry
Applications Immunofluorescence
Applications Immunoprecipitation
Applications Western Blotting
Applications Flow Cytometry
Applications Immunofluorescence
Applications Immunoprecipitation
Applications Western Blotting
Applications Flow Cytometry
Applications Immunofluorescence
Applications Immunoprecipitation
Applications Western Blotting
Classification Monoclonal
Conjugate Unlabeled
Cross Reactivity Bovine
Cross Reactivity Canine
Cross Reactivity Equine
Cross Reactivity Human
Cross Reactivity Porcine
Cross Reactivity Rat
Cross Reactivity Sheep
Cross Reactivity Bovine
Cross Reactivity Canine
Cross Reactivity Equine
Cross Reactivity Human
Cross Reactivity Porcine
Cross Reactivity Rat
Cross Reactivity Sheep
Cross Reactivity Bovine
Cross Reactivity Canine
Cross Reactivity Equine
Cross Reactivity Human
Cross Reactivity Porcine
Cross Reactivity Rat
Cross Reactivity Sheep
Cross Reactivity Bovine
Cross Reactivity Canine
Cross Reactivity Equine
Cross Reactivity Human
Cross Reactivity Porcine
Cross Reactivity Rat
Cross Reactivity Sheep
Cross Reactivity Bovine
Cross Reactivity Canine
Cross Reactivity Equine
Cross Reactivity Human
Cross Reactivity Porcine
Cross Reactivity Rat
Cross Reactivity Sheep
Cross Reactivity Bovine
Cross Reactivity Canine
Cross Reactivity Equine
Cross Reactivity Human
Cross Reactivity Porcine
Cross Reactivity Rat
Cross Reactivity Sheep
Cross Reactivity Bovine
Cross Reactivity Canine
Cross Reactivity Equine
Cross Reactivity Human
Cross Reactivity Porcine
Cross Reactivity Rat
Cross Reactivity Sheep
Format Concentrated
Host Species Mouse
Regulatory Status RUO
Research Discipline Angiogenesis
Type Primary
Immunogen Ca2plus-replete, native purified human TSP from supernatant of thrombin-activated platelets
Isotype IgG1

Provide accurate, reproducible results with the Thermo Scientific™ Thrombospondin (TSP) Ab-3, Mouse Monoclonal Antibody.

Thrombospondin is a protein from platelet a-granules. It is made up of three identical subunits bound by interchain disulfides. It is secreted at sites of platelet activation and aggregation and is involved in the processes of chemotaxis, adhesion, proliferation and differentiation of leukocytes, fibroblasts, smooth muscle and endothelial cells.

Host Species: Mouse

Clone: C6.7

Isotype: IgG1

Species Reactivity: Human, Cow, Pig, Horse, Dog, Sheep and Rat. Others not known.

Epitope: Platelet- or cell-binding domain at the extreme C-terminus of TSP

Immunogen: Ca2+-replete, native purified human TSP from supernatant of thrombin-activated platelets

Molecular Weight: ∽450kDa (Nonreduced)

Cellular Localization: Secretory granules, Golgi complex, endoplasmic reticulum, extracellular matrix Ab-3 shows no cross-reaction with fibronectin, fibrinogen, and von Willebrand factor. It blocks TSP-mediated agglutination of fixed RBCs. It shows no effect on TSP-mediated agglutination of fixed, activated platelets. It inhibits both thrombin- and A23187-induced aggregation of washed, live (not fixed) platelets without affecting the secretion of serotonin. It inhibits adhesion of melanoma G361 cells to TSP. It blocks binding of C-terminal domain to Integrin-Associated Protein (IAP)/CD47.

Recommended for:

  • Immunohistochemistry (Not suitable)
  • Western Blotting (Nonreduced only)
  • Immunofluorescence
  • Immunoprecipitation (Native verified)
  • Electron Microscopy
  • Flow Cytometry
  • Blocks Binding of TSP-1 to IAP/CD47

Note:

USA: RUO; Int'l: RUO