missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Thrombospondin (TSP) Ab-11, Mouse Monoclonal Antibody

Provide accurate, reproducible results with the Thermo Scientific™ Thrombospondin (TSP) Ab-11, Mouse Monoclonal Antibody.

$315.35 - $1,520.10

Specifications

Antigen Thrombospondin (TSP) Ab-11
Clone D4.6 + A6.1 + MBC 200.1
Applications Western Blot
Classification Monoclonal
Conjugate Unlabeled
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Concentration Quantity Price Quantity  
Catalog Number Mfr. No. Concentration Quantity Price Quantity  
MS1066P Thermo Scientific™ Lab Vision™
MS1066P
200μg/mL 1mL Each for $1,431.88
MS1066P0 Thermo Scientific™ Lab Vision™
MS1066P0
200μg/mL 100μL Each for $315.35
MS1066P1 Thermo Scientific
MS1066P1
200μg/mL 500μL Each for $857.55
MS1066P1ABX Thermo Scientific™ Lab Vision™
MS1066P1ABX
1mg/mL 100μL Each for $967.47
MS1066PABX Thermo Scientific™ Lab Vision™
MS1066PABX
1mg/mL 200μL Each for $1,520.10
Specifications

Description

Provide accurate, reproducible results with the Thermo Scientific™ Thrombospondin (TSP) Ab-11, Mouse Monoclonal Antibody.

Thrombospondin is a protein from platelet a-granules. It is made up of three identical subunits bound by interchain disulfides. It is secreted at sites of platelet activation and aggregation and is involved in the processes of chemotaxis, adhesion, proliferation and differentiation of leukocytes, fibroblasts, smooth muscle and endothelial cells.

Host Species: Mouse

Clone: D4.6 + A6.1 + MBC 200.1

Isotype: IgG1 + IgG1 + IgG1

Species Reactivity: Human. Others not known.

Immunogen: Purified human TSP

Molecular Weight: ∽450kDa (nonreduced), 170 to 180kDa (reduced)

Cellular Localization: Secretory granules, Golgi complex, endoplasmic reticulum, extracellular matrix

Recommended for:

  • Immunohistochemistry (Not suitable)
  • Western Blotting (Reduced and nonreduced)

Note:

USA: RUO; Int'l: RUO

Description & Specifications

Specifications

Thrombospondin (TSP) Ab-11
Western Blot
Unlabeled
RUO
Primary
IgG1
D4.6 + A6.1 + MBC 200.1
Monoclonal
Murine
Angiogenesis
Purified human TSP