Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Thrombospondin (TSP) Ab-11, Mouse Monoclonal Antibody


Provide accurate, reproducible results with the Thermo Scientific™ Thrombospondin (TSP) Ab-11, Mouse Monoclonal Antibody.

Manufacturer: thermo scientific™ lab vision™  MS1066PABX

 View more versions of this product

Catalog No. MS1066PABX

  • / Each
 

Description & Specifications

Specifications

Antigen Thrombospondin (TSP) Ab-11
Applications Western Blot
Classification Monoclonal
Clone D4.6 + A6.1 + MBC 200.1
Concentration 1mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Concentrated
Host Species Murine
Immunogen Purified human TSP
Isotype IgG1
Quantity 200μL
Regulatory Status RUO
Research Discipline Angiogenesis
Type Primary
Monoclonal or Polyclonal Monoclonal
Target Species Human

Provide accurate, reproducible results with the Thermo Scientific™ Thrombospondin (TSP) Ab-11, Mouse Monoclonal Antibody.

Thrombospondin is a protein from platelet a-granules. It is made up of three identical subunits bound by interchain disulfides. It is secreted at sites of platelet activation and aggregation and is involved in the processes of chemotaxis, adhesion, proliferation and differentiation of leukocytes, fibroblasts, smooth muscle and endothelial cells.

Host Species: Mouse

Clone: D4.6 + A6.1 + MBC 200.1

Isotype: IgG1 + IgG1 + IgG1

Species Reactivity: Human. Others not known.

Immunogen: Purified human TSP

Molecular Weight: ∽450kDa (nonreduced), 170 to 180kDa (reduced)

Cellular Localization: Secretory granules, Golgi complex, endoplasmic reticulum, extracellular matrix

Recommended for:

  • Immunohistochemistry (Not suitable)
  • Western Blotting (Reduced and nonreduced)

Note:

USA: RUO; Int'l: RUO