Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Thrombospondin (TSP) Ab-11, Mouse Monoclonal Antibody


Provide accurate, reproducible results with the Thermo Scientific™ Thrombospondin (TSP) Ab-11, Mouse Monoclonal Antibody.

Manufacturer: thermo scientific™ lab vision™  MS1066P1ABX

 View more versions of this product

Catalog No. MS1066P1ABX

  • / Each
 

Description & Specifications

Specifications

Regulatory Status RUO
Antigen Thrombospondin (TSP) Ab-11
Host Species Mouse
Isotype IgG1
Format Concentrated
Quantity 100µL
Cross Reactivity Human
Clone D4.6 + A6.1 + MBC 200.1
Immunogen Purified human TSP
Applications Western Blotting
Conjugate Unlabeled
Concentration 1mg/mL
Research Discipline Angiogenesis
Type Primary
Classification Monoclonal

Provide accurate, reproducible results with the Thermo Scientific™ Thrombospondin (TSP) Ab-11, Mouse Monoclonal Antibody.

Thrombospondin is a protein from platelet a-granules. It is made up of three identical subunits bound by interchain disulfides. It is secreted at sites of platelet activation and aggregation and is involved in the processes of chemotaxis, adhesion, proliferation and differentiation of leukocytes, fibroblasts, smooth muscle and endothelial cells.

Host Species: Mouse

Clone: D4.6 + A6.1 + MBC 200.1

Isotype: IgG1 + IgG1 + IgG1

Species Reactivity: Human. Others not known.

Immunogen: Purified human TSP

Molecular Weight: ∽450kDa (nonreduced), 170 to 180kDa (reduced)

Cellular Localization: Secretory granules, Golgi complex, endoplasmic reticulum, extracellular matrix

Recommended for:

  • Immunohistochemistry (Not suitable)
  • Western Blotting (Reduced and nonreduced)

Note:

USA: RUO; Int'l: RUO