missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Thrombospondin (TSP) Ab-1, Mouse Monoclonal Antibody

Provide accurate, reproducible results with the Thermo Scientific™ Thrombospondin (TSP) Ab-1, Mouse Monoclonal Antibody.

$337.55 - $1,718.39

Specifications

Antigen Thrombospondin (TSP) Ab-1
Clone A4.1
Applications Flow Cytometry, Immunofluorescence, Western Blot
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 5
Catalog Number Mfr. No. Concentration Quantity Price Quantity  
Catalog Number Mfr. No. Concentration Quantity Price Quantity  
MS418P View Documents Thermo Scientific™
MS418P
200μg/mL 1mL Each for $1,562.92
 
MS418P0 View Documents Thermo Scientific™
MS418P0
200μg/mL 100μL Each for $337.55
 
MS418P1 View Documents Thermo Scientific™
MS418P1
200μg/mL 500μL Each for $949.73
 
MS418P1ABX View Documents Thermo Scientific™
MS418P1ABX
1mg/mL 100μL Each for $1,055.31
 
MS418PABX View Documents Thermo Scientific™
MS418PABX
1mg/mL 200μL Each for $1,718.39
 
Description

Description

Provide accurate, reproducible results with the Thermo Scientific™ Thrombospondin (TSP) Ab-1, Mouse Monoclonal Antibody.

Thrombospondin is a protein from platelet a-granules. It is made up of three identical subunits bound by interchain disulfides. It is secreted at sites of platelet activation and aggregation and is involved in the processes of chemotaxis, adhesion, proliferation and differentiation of leukocytes, fibroblasts, smooth muscle and endothelial cells.

Host Species: Mouse

Clone: A4.1

Isotype: IgM

Species Reactivity: Human, Mouse, Rat, Cow and Chicken. Others not known.

Epitope: N-terminal half of the central stalk-like region of TSP, which is recovered as a 50kDa fragment after chymotryptic digestion of TSP

Immunogen: Calcium-replete, native purified human TSP from the supernatant of thrombin-activated platelets

Molecular Weight: ∽450kDa (nonreduced)

Positive Control: HUVEC cells

Cellular Localization: Secretory granules, Golgi complex, endoplasmic reticulum, extracellular matrix

Recommended for:

  • Immunohistochemistry (Not suitable)
  • Western Blotting (Nonreduced only)
  • Immunofluorescence
  • Blocks Binding of TSP-1 to TSP-Rec./CD36
  • Flow Cytometry

Note:

USA: RUO; Int'l: RUO

General References:

1. Prater CA, et. al. J Cell Biol, 1991, 112(5):1031-40.

Specifications

Specifications

Thrombospondin (TSP) Ab-1
Flow Cytometry, Immunofluorescence, Western Blot
Unconjugated
Murine
Angiogenesis
Calcium-replete, native purified human TSP from the supernatant of thrombin-activated platelets
A4.1
Monoclonal
Concentrated
RUO
Primary
IgM