missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Thrombospondin-1 (TSP-1) Ab-2, Mouse Monoclonal Antibody

Provide accurate, reproducible results with the Thermo Scientific™ Thrombospondin-1 (TSP-1) Ab-2, Mouse Monoclonal Antibody.

$904.98 - $904.98

Specifications

Antigen Thrombospondin-1 (TSP-1) Ab-2
Clone D4.6
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Format Concentrated
View More Specs

Products 1
Catalog Number Mfr. No. Concentration Conjugate Quantity Price Quantity  
Catalog Number Mfr. No. Concentration Conjugate Quantity Price Quantity  
MS419P1 View Documents Thermo Scientific™
MS419P1
200μg/mL Unconjugated 500μL Each for $904.98
 
Description

Description

Provide accurate, reproducible results with the Thermo Scientific™ Thrombospondin-1 (TSP-1) Ab-2, Mouse Monoclonal Antibody.

Thrombospondin is a protein from platelet a-granules. It is made up of three identical subunits bound by interchain disulfides. It is secreted at sites of platelet activation and aggregation and is involved in the processes of chemotaxis, adhesion, proliferation and differentiation of leukocytes, fibroblasts, smooth muscle and endothelial cells.

Host Species: Mouse

Clone: D4.6

Isotype: IgG1

Species Reactivity: Human, Mouse, Rat, Cow, Pig, Horse, Dog and Sheep. Others not known.

Epitope: Calcium-binding domain of TSP (C-terminal 50kDa piece of the 120kDa fragment)

Immunogen: Reduced and alkylated purified human TSP (fully denatured) from the supernatant of thrombin-activated platelets

Molecular Weight: ∽450kDa (nonreduced), 170 to 180kDa (reduced)

Cellular Localization: Secretory granules, Golgi complex, endoplasmic reticulum, extracellular matrix

Recommended for:

  • Immunohistochemistry (Not suitable)
  • Western Blotting
  • Immunoprecipitation (Native verified)
  • Electron Microscopy

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

Thrombospondin-1 (TSP-1) Ab-2
Immunoprecipitation, Western Blot
Concentrated
RUO
Primary
IgG1
D4.6
Monoclonal
Murine
Angiogenesis
Reduced and alkylated purified human TSP (fully denatured) from the supernatant of thrombin-activated platelets