Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Thrombospondin-1 (TSP-1) Ab-2, Mouse Monoclonal Antibody


Provide accurate, reproducible results with the Thermo Scientific™ Thrombospondin-1 (TSP-1) Ab-2, Mouse Monoclonal Antibody.

Manufacturer: thermo scientific™ lab vision™  MS419P1ABX

 View more versions of this product

Catalog No. MS419P1ABX

  • / Each
 

Description & Specifications

Specifications

Applications Immunoprecipitation
Applications Western Blotting
Quantity 100µL
Format Concentrated
Isotype IgG1
Cross Reactivity Bovine
Cross Reactivity Canine
Cross Reactivity Equine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Porcine
Cross Reactivity Rat
Cross Reactivity Sheep
Host Species Mouse
Antigen Thrombospondin-1 (TSP-1) Ab-2
Immunogen Reduced and alkylated purified human TSP (fully denatured) from the supernatant of thrombin-activated platelets
Clone D4.6
Regulatory Status RUO
Type Primary
Research Discipline Angiogenesis
Classification Monoclonal
Concentration 1mg/mL
Conjugate Unlabeled

Provide accurate, reproducible results with the Thermo Scientific™ Thrombospondin-1 (TSP-1) Ab-2, Mouse Monoclonal Antibody.

Thrombospondin is a protein from platelet a-granules. It is made up of three identical subunits bound by interchain disulfides. It is secreted at sites of platelet activation and aggregation and is involved in the processes of chemotaxis, adhesion, proliferation and differentiation of leukocytes, fibroblasts, smooth muscle and endothelial cells.

Host Species: Mouse

Clone: D4.6

Isotype: IgG1

Species Reactivity: Human, Mouse, Rat, Cow, Pig, Horse, Dog and Sheep. Others not known.

Epitope: Calcium-binding domain of TSP (C-terminal 50kDa piece of the 120kDa fragment)

Immunogen: Reduced and alkylated purified human TSP (fully denatured) from the supernatant of thrombin-activated platelets

Molecular Weight: ∽450kDa (nonreduced), 170 to 180kDa (reduced)

Cellular Localization: Secretory granules, Golgi complex, endoplasmic reticulum, extracellular matrix

Recommended for:

  • Immunohistochemistry (Not suitable)
  • Western Blotting
  • Immunoprecipitation (Native verified)
  • Electron Microscopy

Note:

USA: RUO; Int'l: RUO