Please login to your online account to display your discounted pricing

Thrombospondin-1 Mouse, Clone: A6.1, eBioscience™

Mouse Monoclonal Antibody

$160.95 - $397.30

Specifications

Antigen Thrombospondin-1
Clone A6.1
Host Species Murine
Species Reactivity Human, Mouse, Rat
Applications ELISA
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501122825

 
life technologies
LS14975680
25μg Each for $160.95

501122826

 
life technologies
LS14975682
100μg Each for $397.30
Description & Specifications

Specifications

Antigen Thrombospondin-1
Clone A6.1
Host Species Murine
Species Reactivity Human, Mouse, Rat
Applications ELISA
Applications Immunocytochemistry
Applications Immunohistochemistry (Formalin/Paraffin)
Applications Immunoprecipitation
Applications Microscopy
Applications Western Blot
Regulatory Status RUO
Conjugate Unlabeled
Format Purified
Storage Requirements Store at 2-8°C.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.5mg/mL

The A6.1 monoclonal antibody recognizes human, mouse, and rat thrombospondin-1, a member of the thrombospondin family of extracellular calcium-binding proteins. Thrombospondin-1 is a secreted glycoprotein that was first discovered in activated platelets but is also expressed by endothelial cells, adipocytes, fibroblasts, macrophages, monocytes, and smooth muscle cells. Thrombospondin-1 functions by binding members of the extracellular matrix such as fibronectin, integrins, and heparin, and its signaling is mediated through activation of the TGF beta signaling pathway. Thrombospondin-1 has been shown to be involved in angiogenesis, inflammation, osteogenesis, cell proliferation, and synaptogenesis during neural development.