Please login to your online account to display your discounted pricing

Alfa Aesar™ Thrombin Receptor Activator for Peptide 5

For cellular and molecular biology applications

Specifications

Product Type Thrombin Receptor Activator for Peptide 5
Molecular Weight 634.80
Storage Requirements -30° to -10°C
Format Solid
Sequence Ser-Phe-Leu-Leu-Arg
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66983MCR

 
alfa aesar™
J66983MCR
1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Product Type Thrombin Receptor Activator for Peptide 5
Molecular Weight 634.80
Storage Requirements -30° to -10°C
Format Solid
Sequence Ser-Phe-Leu-Leu-Arg
Common Name TRAP-5
Regulatory Status RUO
Purification Method HPLC