Please login to your online account to display your discounted pricing

Alfa Aesar™ Thrombin Receptor Activator for Peptide 5

For cellular and molecular biology applications

Manufacturer: alfa aesar™  J66983EXD

 View all options for this product

Catalog No. AAJ66983EXD

 Please call Customer Service at 1-800-234-7437 or send an email to for assistance.

Description & Specifications


Molecular Weight 634.80
Purification Method HPLC
Quantity 0.5mg
Format Solid
Sequence Ser-Phe-Leu-Leu-Arg
Regulatory Status RUO
Storage Requirements -30° to -10°C
Common Name TRAP-5
Product Type Thrombin Receptor Activator for Peptide 5