Please login to your online account to display your discounted pricing

Alfa Aesar™ Thrombin Receptor Activator for Peptide 5


For cellular and molecular biology applications

Manufacturer: alfa aesar™  J66983MCR

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. AAJ66983MCR

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Specifications

Storage Requirements -30° to -10°C
Format Solid
Molecular Weight 634.80
Sequence Ser-Phe-Leu-Leu-Arg
Common Name TRAP-5
Purification Method HPLC
Product Type Thrombin Receptor Activator for Peptide 5
Regulatory Status RUO
Quantity 1mg