Thermo Scientific Chemicals Thrombin, Bovine Plasma
For research use
Supplier:  Thermo Scientific Chemicals J63383EXP
 View more versions of this product
Catalog No. AAJ63383EXP
Description
Description
Thrombin is a trypsin-like serine protease involved in a multitude of processes in the human body. Thrombin generation is the result of limited proteolysis of the vitamin K-dependent zymogen prothrombin. Thrombin is the last enzyme in the clotting cascade functioning to cleave fibrinogen to fibrin which forms the fibrin gel of a haemostatic plug or a pathologic thrombus
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Lyophilized powder, Reconstitute in 0.9% sodium chloride
Bovine plasma
RUO
Native
100KU
-30° to -10°C
Thrombin, Bovine Plasma
Lyophilized