Please login to your online account to display your discounted pricing

threo Ifenprodil hemitartrate, Tocris

NR2B-selective NMDA antagonist; Also σ agonist

$205.70 - $862.95

Chemical Identifiers

Chemical Name or Material threo Ifenprodil hemitartrate
Assay Percent Range >98%
Molecular Formula C21H27NO2·+C4H6O6
Formula Weight 400.49
Synonym (1S™, 2S™)-threo-2-(4-Benzylpiperidino)-1-(4-hydroxyphenyl)-1-propanol hemitartrate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

289250

 
r & d systems
RDS289250
50mg Each for $862.95

289210

 
r & d systems
RDS289210
10mg Each for $205.70
Description & Specifications

Specifications

Formula Weight 400.49
Quantity 50mg
Chemical Name or Material threo Ifenprodil hemitartrate
Assay Percent Range >98%
Molecular Formula C21H27NO2·+C4H6O6
CAS 1312991-83-5
Synonym (1S™, 2S™)-threo-2-(4-Benzylpiperidino)-1-(4-hydroxyphenyl)-1-propanol hemitartrate
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature