Please login to your online account to display your discounted pricing

(±)-threo-3-Methylglutamic acid, Tocris


EAAT2 and EAAT4 blocker

Quantity 10mg

Manufacturer: r & d systems  RDS081110

 View more versions of this product

Catalog No. 081110

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 10mg
Formula Weight 161.16
CAS 63088-04-0
Chemical Name or Material (±)-threo-3-Methylglutamic acid
Recommended Storage Store at Room Temperature
Assay Percent Range >98%
Molecular Formula C6H11NO4

Specifications

Chemical Name or Material (±)-threo-3-Methylglutamic acid
Assay Percent Range >98%
Molecular Formula C6H11NO4
Formula Weight 161.16
Accessories

Store at Room Temperature