Please login to your online account to display your discounted pricing

(±)-threo-3-Methylglutamic acid, Tocris

EAAT2 and EAAT4 blocker

$146.74 - $618.87

Chemical Identifiers

Chemical Name or Material (±)-threo-3-Methylglutamic acid
Assay Percent Range >98%
Molecular Formula C6H11NO4
Formula Weight 161.16
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

081110

 
r & d systems
RDS081110
10mg Each for $146.74

081150

 
r & d systems
RDS081150
50mg Each for $618.87
Description & Specifications

Specifications

Quantity 10mg
Formula Weight 161.16
CAS 63088-04-0
Chemical Name or Material (±)-threo-3-Methylglutamic acid
Recommended Storage Store at Room Temperature
Assay Percent Range >98%
Molecular Formula C6H11NO4
Safety and Handling

Store at Room Temperature