Please login to your online account to display your discounted pricing

Alfa Aesar™ [Thr&superscript_4;,Gly&superscript_7;] Oxytocin

For cellular and molecular biology applications

$68.35 - $238.59

Specifications

Product Type [Thr4,Gly7] Oxytocin
Molecular Weight 940.10
Storage Requirements -30° to -10°C
Sequence Cys-Tyr-Ile-Thr-Asn-Cys-Gly-Leu-Gly-NH2 (Disulfide bridge: Cys1-Cys6)
Format Solid
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66756LB0

 
alfa aesar™
J66756LB0
5mg Each for $238.59

AAJ66756MCR

 
alfa aesar™
J66756MCR
1mg Each for $68.35
Description & Specifications

Specifications

Product Type [Thr4,Gly7] Oxytocin
Molecular Weight 940.10
Storage Requirements -30° to -10°C
Sequence Cys-Tyr-Ile-Thr-Asn-Cys-Gly-Leu-Gly-NH2 (Disulfide bridge: Cys1-Cys6)
Format Solid
Regulatory Status RUO
Purification Method HPLC