Please login to your online account to display your discounted pricing

Alfa Aesar™ [Thr&superscript_4;,Gly&superscript_7;] Oxytocin


For cellular and molecular biology applications

Manufacturer: alfa aesar™  J66756MCR

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. AAJ66756MCR

  • / Each
 

Description & Specifications

Specifications

Quantity 1mg
Format Solid
Sequence Cys-Tyr-Ile-Thr-Asn-Cys-Gly-Leu-Gly-NH2 (Disulfide bridge: Cys1-Cys6)
Regulatory Status RUO
Storage Requirements -30° to -10°C
Product Type [Thr4,Gly7] Oxytocin
Molecular Weight 940.10
Purification Method HPLC