Please login to your online account to display your discounted pricing

Alfa Aesar™ [Thr&superscript_begin;28&superscript_end;, Nle&superscript_begin;31&superscript_end;] Cholecystokinin (25-33), Sulfated

For cellular and molecular biology applications

Specifications

Product Type [Thr28, Nle31] Cholecystokinin (25-33), sulfated
Molecular Weight 1251.32
Storage Requirements -30° to -10°C
Sequence Arg-Asp-Tyr(SO3H)-Thr-Gly-Trp-Nle-Asp-Phe-NH2
Format Solid
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66253EXD

 
alfa aesar™
J66253EXD
0.5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ66253MCR

 
alfa aesar™
J66253MCR
1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Product Type [Thr28, Nle31] Cholecystokinin (25-33), sulfated
Molecular Weight 1251.32
Storage Requirements -30° to -10°C
Sequence Arg-Asp-Tyr(SO3H)-Thr-Gly-Trp-Nle-Asp-Phe-NH2
Format Solid
Regulatory Status RUO
Purification Method HPLC