Please login to your online account to display your discounted pricing

ThPOK Rat anti-Mouse, PE, Clone: 2POK, eBioscience™

Rat Monoclonal Antibody

$172.55 - $433.55

Specifications

Antigen ThPOK
Clone 2POK
Host Species Rat
Gene Alias c-Krox, Zbtb7b
Species Reactivity Mouse
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501122354

 
life technologies
LS12592880
25μg Each for $172.55

501122355

 
life technologies
LS12592882
100μg Each for $433.55
Description & Specifications

Specifications

Antigen ThPOK
Clone 2POK
Host Species Rat
Gene Alias c-Krox, Zbtb7b
Species Reactivity Mouse
Applications Flow Cytometry (Intracellular Staining)
Regulatory Status RUO
Conjugate PE
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL

This 2POK monoclonal antibody reacts with mouse ThPOK, which is also known as c-Krox and Zbtb7b. In hematopoietic cells, this 78kDa transcription factor is expressed in thymocytes and T lymphocytes in the spleen and lymph nodes. In non-hematopoietic cells, ThPOK can be detected in the skin, heart, smooth muscle, and liver. Studies demonstrate that ThPOK promotes CD4+ lineage differentiation by inhibiting the expression of CD8 lineage-specific genes such as Runx3. ThPOK has also been shown to inhibit Cd4 silencing, another key facet required for differentiation to the CD4+ lineage. In addition to its role in thymocyte development, ThPOK is involved in the effector functions of invariant natural killer T (iNKT) cells and gamma delta T cells.