missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thorium AA Standard, 1mL = 1mg Th (1,000ppm Th)Th(NO3)4 in 3% HNO3, Ricca Chemical

Chemical Identifiers

CAS 7732-18-5,7697-37-2,13823-29-5
CAS Min % 94.0,4.22,0.23
CAS Max % 97.0,4.39,0.23
Chemical Name or Material Thorium AA Standard
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
ATH1KN100 Ricca Chemical Company
RIATH1KN100
Poly natural 100mL N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
ATH1KN500 Ricca Chemical Company
RIATH1KN500
Poly natural 500mL N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

7732-18-5,7697-37-2,13823-29-5
94.0,4.22,0.23
97.0,4.39,0.23
Thorium AA Standard

Specifications

7732-18-5,7697-37-2,13823-29-5
97.0,4.39,0.23
Odorless
Clear colorless
Thorium AA Standard
94.0,4.22,0.23
1mL = 1mg Th (1, 000ppm Th)Th(NO3)4 in 3% HNO3
Poly natural
100mL

Safety and Handling

ShelfLife : 24 months