Please login to your online account to display your discounted pricing

Alfa Aesar™ Thorin

Chemical Identifiers

Chemical Name or Material Thorin
CAS 3688-92-4
Molecular Formula C16H11AsN2Na2O10S2
Formula Weight 576.3
MDL Number MFCD00003888
UN Number UN3465
Beilstein 2957648
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1029706

 
alfa aesar™
L1029706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1029714

 
alfa aesar™
L1029714
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 576.3
TSCA TSCA
Molecular Formula C16H11AsN2Na2O10S2
CAS 3688-92-4
Health Hazard 1 Danger
MDL Number MFCD00003888
UN Number UN3465
Beilstein 2957648
Health Hazard 2 GHS H Statement
H301-H331
Toxic if swallowed.
Toxic if inhaled.
Health Hazard 3 GHS P Statement
P261-P301+P310-P304+P340-P311-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ORGANOARSENIC COMPOUND, SOLID, N.O.S.
Melting Point >300°C
Quantity 5g
Chemical Name or Material Thorin

Applications
Thorin is used as a reagent for the determination of thorium. It is also used as an indicator in the titrimetric determination of organic sulfur compounds and sulfate with Ba(ClO4)2.

Solubility
Soluble in water.

Notes
Stable under recommended storage conditions. Store away from oxidizing agents.

Safety and Handling

Danger

GHS P Statement
P261-P301+P310-P304+P340-P311-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H331
Toxic if swallowed.
Toxic if inhaled.