Please login to your online account to display your discounted pricing

Alfa Aesar™ Thorin


Quantity 25g

Manufacturer: alfa aesar™  L1029714

 View more versions of this product

Catalog No. AAL1029714

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 576.3
Chemical Name or Material Thorin
TSCA TSCA
Molecular Formula C16H11AsN2Na2O10S2
CAS 3688-92-4
Beilstein 2957648
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement
P261-P301+P310-P304+P340-P311-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 2 GHS H Statement
H301-H331
Toxic if swallowed.
Toxic if inhaled.
DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ORGANOARSENIC COMPOUND, SOLID, N.O.S.
MDL Number MFCD00003888
UN Number UN3465
Melting Point >300°C
Quantity 25g

Specifications

Chemical Name or Material Thorin
CAS 3688-92-4
Molecular Formula C16H11AsN2Na2O10S2
Formula Weight 576.3
MDL Number MFCD00003888
UN Number UN3465
Beilstein 2957648

Applications
Thorin is used as a reagent for the determination of thorium. It is also used as an indicator in the titrimetric determination of organic sulfur compounds and sulfate with Ba(ClO4)2.

Solubility
Soluble in water.

Notes
Stable under recommended storage conditions. Store away from oxidizing agents.

Accessories

Danger

GHS P Statement
P261-P301+P310-P304+P340-P311-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H331
Toxic if swallowed.
Toxic if inhaled.