missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Thomsen-Friedenreich Antigen Ab-1, Mouse Monoclonal Antibody

Provide accurate, reproducible results with the Thermo Scientific™ Thomsen-Friedenreich Antigen Ab-1, Mouse Monoclonal Antibody.

Manufacturer:  Thermo Scientific™ Lab Vision™ MS455P1ABX

 View more versions of this product

Catalog No. MS455P1ABX


 

Specifications

Description

Provide accurate, reproducible results with the Thermo Scientific™ Thomsen-Friedenreich Antigen Ab-1, Mouse Monoclonal Antibody.

The Thomsen-Fridenreich antigen acts as an oncofetal antigen, with low expression in normal adult tissues but increasing to fetal levels of expression in hyperplasia or malignancy. It is considered a pan-carcinoma marker.

Host Species: Mouse

Clone: A78-G/A7

Isotype: IgM/κ

Species Reactivity: Human and Rat. Others not known.

Epitope: Galβ1-3GalNAc

Immunogen: Neuraminidase-treated human red blood cells

Positive Control: KG1 cells, Human colorectal CA tissues

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Immunofluorescence

Note:

USA: RUO; Int'l: RUO

Description & Specifications

Specifications

Thomsen-Friedenreich Antigen Ab-1
Monoclonal
1mg/mL
Ab without BSA/Azide (Frozen)
Murine
IgM kappa
RUO
Primary
Human,Rat
Immunofluorescence,Immunohistochemistry (Paraffin)
A78-G/A7
Unlabeled
Concentrated
Neuraminidase-treated human red blood cells
100μL
Cancer and Tumor Biology
Monoclonal