Please login to your online account to display your discounted pricing

Alfa Aesar™ Thioxanthen-9-one, 98%

$189.34 - $189.34

Chemical Identifiers

Chemical Name or Material Thioxanthen-9-one
CAS 492-22-8
Molecular Formula C13H8OS
Formula Weight 212.27
MDL Number MFCD00005066
Beilstein 140978
Merck Index 14,9369
Synonym Thioxanthone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2376422

 
alfa aesar™
B2376422
100g Each for $189.34
Description & Specifications

Specifications

Formula Weight 212.27
Chemical Name or Material Thioxanthen-9-one
TSCA TSCA
Assay 98%
Molecular Formula C13H8OS
MDL Number MFCD00005066
CAS 492-22-8
Beilstein 140978
Merck Index 14,9369
Synonym Thioxanthone
Melting Point 216° to 218°C
Quantity 100g