Learn More
Thiourea, ACS, 99% min., Thermo Scientific Chemicals
Reagent for organic synthesis
Supplier: Thermo Scientific Chemicals 03660930
Description
Used as a reagent for organic synthesis. Thiourea is a photographic fixative, and used in manufacture of resins. It act as a catalyst for asymmetric reactions. It plays an essential role as catalyst for highly enantio- and diastereoselective additions reaction of oxindoles to nitroolefins. It is also useful to improve the productivity of mung bean.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Thiourea | |
175°C to 178°C | |
1.405 g/mL | |
CH4N2S | |
MFCD00008067 | |
UN3077 | |
14,9367 | |
Soluble in water. | |
NC(N)=S | |
76.12 | |
CHEBI:36946 | |
≤99% | |
Crystalline |
62-56-6 | |
White | |
Odorless | |
H2NCSNH2 | |
250 g | |
605327 | |
thiocarbamide, 2-thiourea, pseudothiourea, isothiourea, sulourea, 2-thiopseudourea, sulfourea, thiuronium, thiocarbonic acid diamide, urea, thio | |
UMGDCJDMYOKAJW-UHFFFAOYSA-N | |
thiourea | |
2723790 | |
76.12 | |
ACS |
Chemical Identifiers
62-56-6 | |
76.12 | |
UMGDCJDMYOKAJW-UHFFFAOYSA-N | |
2723790 | |
thiourea |
CH4N2S | |
MFCD00008067 | |
thiocarbamide, 2-thiourea, pseudothiourea, isothiourea, sulourea, 2-thiopseudourea, sulfourea, thiuronium, thiocarbonic acid diamide, urea, thio | |
CHEBI:36946 | |
NC(N)=S |
Safety and Handling
GHS H Statement
H351-H361-H302
Suspected of causing cancer.
Suspected of damaging fertility or the unborn child.
Harmful if swallowed.
P201-P202-P264b-P270-P281-P301+P312-P308+P313-P330-P501c
H302-H350-H361d-H500
DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber : 200-543-5
RTECSNumber : YU2800000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only