missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thiourea, 99+%, ACS reagent, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 424540010

 View more versions of this product

Catalog No. AC424540010



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Thiourea
62-56-6
White
0.5% max.
99% min. (On dried basis) (Argentometry)
CH4N2S
MFCD00008067
03, 180
(to bismuth) Passes Test
thiocarbamide, 2-thiourea, pseudothiourea, isothiourea, sulourea, 2-thiopseudourea, sulfourea, thiuronium, thiocarbonic acid diamide, urea, thio
UMGDCJDMYOKAJW-UHFFFAOYSA-N
thiourea
2723790
76.12
ACS Reagent
99+%
171.0°C to 175.0°C
0.1% max.
Authentic
Plastic bottle
H2NCSNH2
1 kg
01,1164; 02,412; 03,290; 06,586; 11,519; 15,308
15, 9521
(16.5g in 100ml water,25°C) Passes test
NC(N)=S
76.12
CHEBI:36946
99+%
Crystalline Powder

Chemical Identifiers

62-56-6
76.12
UMGDCJDMYOKAJW-UHFFFAOYSA-N
2723790
thiourea
CH4N2S
MFCD00008067
thiocarbamide, 2-thiourea, pseudothiourea, isothiourea, sulourea, 2-thiopseudourea, sulfourea, thiuronium, thiocarbonic acid diamide, urea, thio
CHEBI:36946
NC(N)=S
Safety and Handling

Safety and Handling

GHS H Statement
Suspected of causing cancer.
Toxic to aquatic life with long lasting effects.
Suspected of damaging the unborn child.
Harmful if swallowed.

GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Immediately call a POISON CENTER or doctor/physician.
Use personal protective equipment as required.
IF INHALED: Remove victim to fresh air and keep

GHS Signal Word: Warning

EINECSNumber : 200-543-5

RTECSNumber : YU2800000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only